missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RelA/NFkB p65 Rat anti-Human, Alexa Fluor 594, Clone: 465003, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems IC50781T-100UG
This item is not returnable.
View return policy
Description
RelA/NFkB p65 Monoclonal specifically detects RelA/NFkB p65 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| RelA/NFkB p65 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_068810 | |
| RELA | |
| E. coli-derived recombinant human RelA/NF kappa B p65 Met1-Thr254 Accession # NP_068810 | |
| RUO | |
| 5970 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 465003 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, p65RelA, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects human RelA/NF kappa B p65 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only (RUO)