Learn More
Tuttnauer Release Trolley
Product Code. 17848269 Shop All Tuttnauer Products
Click to view available options
:
Release Trolley; For use with: Tiva Washer
Product Code. 17848269
Brand: Tuttnauer ATS2380501
Only null left
Add to basket
Add to basket
Release Trolley suitable for the transport of washing trolley. Loading trolley with block system to load washing material into the washer disinfector. Anti-drop system to collect sinking water from load material. Wheels able to be blocked. Anti-upside-down system for security transport.
  • Suitable for the transport of washing trolley.
  • Loading trolley with block system to load washing material into the washer disinfector.
  • Anti-drop system to collect sinking water from load material.
  • Wheels able to be blocked.
  • Anti-upside-down system for security transport.
  • Ergonomic handle and design.
  • W x D x H: 634 x 633 x 880 mm (24.96 x 24.92 x 34.64 in.)
Specifications
Material Stainless Steel
Product Type Trolley
For Use With (Equipment) Tiva Washer
Quantity 1 Ea.