missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H

Product Code. 12677499
Change view
Click to view available options
Description:
Polyvalent, Phases 1 and 2
Rapid Diagnostic 1 (b, d, E, r)
Rapid Diagnostic 2 (b, E, k, L)
Rapid Diagnostic 3 (d, E, G, k)
Salmonella H (1, 2)
Salmonella H (1, 5)
Salmonella H (1, 6)
Salmonella H (1, 7)
Salmonella H (L) Polyvalent
Salmonella H (a)
Salmonella H (b)
Salmonella H (c )
Salmonella H (d)
Salmonella H (eh)
Salmonella H (enx)
Salmonella H (gm)
Salmonella H (r )
Salmonella H (y)
Salmonella H (z27)
Unit Size:
2mL
Each
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
12677499 Polyvalent, Phases 1 and 2 Each
11933342 Salmonella H (c ) 2mL
11983332 Salmonella H (a) 2mL
12697429 Salmonella H (1, 7) Each
12719290 Salmonella H (enx) Each
11923342 Salmonella H (b) 2mL
12789280 Salmonella H (d) Each
12667429 Salmonella H (1, 2) Each
12677429 Salmonella H (1, 5) Each
12687429 Salmonella H (1, 6) Each
12759290 Salmonella H (gm) Each
12759300 Salmonella H (r ) Each
12709290 Salmonella H (eh) Each
12617429 Salmonella H (z27) Each
12749280 Rapid Diagnostic 3 (d, E, G, k) Each
12719300 Salmonella H (L) Polyvalent Each
12660008 Salmonella H (y) Each
12697579 Rapid Diagnostic 1 (b, d, E, r) Each
12623658 Rapid Diagnostic 2 (b, E, k, L) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Product Code. 12677499 Supplier Thermo Scientific™ Supplier No. R30858501
  • ONLINE EXCLUSIVE PRICE
    142.71 €
    132.90 € / Pack of 1
    Save 9.81 €
    6% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.

Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.

  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY

Specifications

Control Sets Positive
Description Polyvalent, Phases 1 and 2
Detectable Analytes Salmonella H Antigen
Type Agglutinating Sera
Material Polyvalent
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella H
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.