missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ BactiDrop™ Ferric Chloride

Product Code. 12719240
Change view
Click to view available options
Quantity:
50 x 0.75 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12719240 50 x 0.75 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12719240 Supplier Thermo Scientific™ Supplier No. R21514
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use BactiDrop™ Ferric Chloride (10%) reagent for detecting phenylalanine deamination in the phenylalanine test.

Detect deamination of phenylalanine using Thermo Scientific™ Remel™ BactiDrop™ Ferric Chloride (10%) reagent. Henecka and Houben-Weyl demonstrated that when testing for the presence of phenylpyruvic acid using an aqueous solution of ferric chloride, a green color reaction is produced1,2. This biochemical test is useful for differentiating Enterobacteriaceae. Most members of the Proteus and Providencia groups give a positive phenylalanine test reaction3,4.

The ability to deaminate phenylalanine to phenylpyruvic acid is measured by the phenylalanine test. Ferric chloride acts as a chelating agent that reacts with phenylpyruvic acid and forms a green color5.Ready-to-use: Liquid reagentRapid test: Visible color change within 5 minutes of addition of reagentEasy to interpret:Positive Test - Green color development within 5 minutesNegative Test - No color developmentBactiDrop™ Ferric Chloride (10%) contains Ferric Chloride,6H2O in hydrochloric acid and demineralized water and is supplied as 0.75mL ampules.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 20°C to 25°C
Description BactiDrop Ferric Chloride
CE Marker Yes
Quantity 50 x 0.75 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.