Learn More
Thermo Scientific™ Remel™ BactiDrop India Ink
Product Code. 12729240
Click to view available options
Quantity:
50 Pk.
Unit Size:
Each
Product Code. 12729240
Brand: Thermo Scientific™ R21518
Remel BactiDrop India Ink is for microscopic examination of encapsulated microorganisms.
Remel BactiDrop India Ink is for microscopic examination of encapsulated microorganisms.
Ordering Alerts: Supplied in crushable ampules enclosed in a protective plastic sheath with a convenient dropper dispenser tip.
TRUSTED_SUSTAINABILITY
Specifications
Type India Ink
Quantity 50 Pk.
Volume (Metric) 0.75 mL/Ampule