missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BactiStaph™ Latex
Descripción
Rapidly identify Staphylococcus aureus and distinguish from other staphylococci by detecting clumping factor and protein A using the Thermo Scientific™ BactiStaph™ Latex.
Use this rapid test utilizing latex particles which are capable of detecting both clumping factor and protein A simultaneously. Agglutination of white latex particles is visible to the unaided eye within 60 seconds and represents a positive reaction.
Use with suspected S. aureus colonies. Prior to testing, suspect colonies should be Gram stained and a catalase test performed to verify that they are Staphylococcus.
Although Staphylococci are commonly found on the skin and in mucous membranes, they have been associated with many human and animal infections. S.aureus and other coagulase-positive Staphylococci have been identified as a cause of suppurative infections, food poisoning and toxic shock syndrome and have been isolated from nearly all anatomical sites.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Use with suspected S. aureus colonies. Prior to testing, suspect colonies should be Gram stained and a catalase test performed to verify that they are Staphylococcus.
Although Staphylococci are commonly found on the skin and in mucous membranes, they have been associated with many human and animal infections. S.aureus and other coagulase-positive Staphylococci have been identified as a cause of suppurative infections, food poisoning and toxic shock syndrome and have been isolated from nearly all anatomical sites.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Especificaciones
Especificaciones
| Content And Storage | 2°C to 8°C |
| Control Sets | Positive,Negative |
| Description | BactiStaph Latex |
| CE Marker | Yes |
| Detectable Analytes | S. aureus Coagulase, Protein A |
| Type | Staphylococcus aureus Test Kit |
| Includes | Latex reagent, positive control, negative control, disposable slides, stirring sticks |
| Material | Latex |
| Quantity | 450 Tests/Kit |
| Target Organism Class | Staphylococcus aureus |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?