missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BactiStaph™ Latex

Product Code. 12619937
Change view
Click to view available options
Quantity:
150 Tests/Kit
450 Tests/Kit
Unit Size:
150 tests
450 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12619937 150 Tests/Kit 150 tests
12609937 450 Tests/Kit 450 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12619937 Supplier Thermo Scientific™ Supplier No. R21144
  • ONLINE EXCLUSIVE PRICE
    379.71 €
    290.40 € / 150 tests
    Save 89.31 €
    23% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect clumping factor and protein A associated with the identification of Staphylococcus aureus while distinguishing S. aureus from other staphylococci.

Rapidly identify Staphylococcus aureus and distinguish from other staphylococci by detecting clumping factor and protein A using the Thermo Scientific™ BactiStaph™ Latex.

Use this rapid test utilizing latex particles which are capable of detecting both clumping factor and protein A simultaneously. Agglutination of white latex particles is visible to the unaided eye within 60 seconds and represents a positive reaction.

Use with suspected S. aureus colonies. Prior to testing, suspect colonies should be Gram stained and a catalase test performed to verify that they are Staphylococcus.

Although Staphylococci are commonly found on the skin and in mucous membranes, they have been associated with many human and animal infections. S.aureus and other coagulase-positive Staphylococci have been identified as a cause of suppurative infections, food poisoning and toxic shock syndrome and have been isolated from nearly all anatomical sites.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets Positive,Negative
Description BactiStaph Latex
CE Marker Yes
Detectable Analytes S. aureus Coagulase, Protein A
Type Staphylococcus aureus Test Kit
Includes Latex reagent, positive control, negative control, disposable slides, stirring sticks
Material Latex
Quantity 150 Tests/Kit
Target Organism Class Staphylococcus aureus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.