missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Coagulase Plasma

Product Code. 11943322
Change view
Click to view available options
Quantity:
5 mL
15 mL
25 mL
Unit Size:
15mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11943322 5 mL 5mL
10220293 25 mL 25mL
10046782 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11943322 Supplier Thermo Scientific™ Supplier No. R21050
  • ONLINE EXCLUSIVE PRICE
    72.90 €
    71.10 € / 5mL
    Save 1.80 €
    2% Off
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Detect coagulase enzyme in staphylococci with Coagulase Plasma.

Detect coagulase enzyme in staphylococci with Thermo Scientific™ Remel™ Coagulase Plasma. The enzyme coagulase acts on a constituent of rabbit plasma to produce a thrombin-like substance which activates fibrinogen to form fibrin clot. In 1940, Fairbrother and Chapman et al. reported pathogenic staphylococci can be identified based on the ability to coagulate plasma.1,2 Chapman found rabbit plasma to be superior to other types of plasma in terms of clotting activity.3 Bayliss and Hall recommended replacing the citrate anticoagulant with EDTA to avoid false-positive clots.4

  • Rabbit plasma with EDTA
  • Lyophilized
TRUSTED_SUSTAINABILITY

Specifications

Description Coagulase Plasma
CE Marker Yes
Detectable Analytes Staphylococcus species
Type Coagulase Plasma
For Use With (Application) Enzyme Immunoassay Detection
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.