missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Gram Crystal Violet

Product Code. 11914092
Click to view available options
Quantity:
5/Pk.
Each
Unit Size:
250mL
4 L
Pack of 5
This item is not returnable. View return policy

Product Code. 11914092

Brand: Thermo Scientific™ R40073

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.

Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.

  • For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
  • Selected Gram stains are CE marked
TRUSTED_SUSTAINABILITY

Specifications

Type Crystal Violet
Quantity Each
Certifications/Compliance CE Marked
Volume (Metric) 3.785 L
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.