missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ PathoDx™ Extraction Reagent 2

Product Code. 10402123
Change view
Click to view available options
Description:
PathoDx Extraction Reagent 2, 14mL/Vial
PathoDx Extraction Reagent 2, 7mL/Vial
Quantity:
Each
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity unitSize
10402123 PathoDx Extraction Reagent 2, 7mL/Vial Each Each
12771770 PathoDx Extraction Reagent 2, 14mL/Vial Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10402123 Supplier Thermo Scientific™ Supplier No. R62055
  • ONLINE EXCLUSIVE PRICE
    40.10 €
    39.07 € / Pack of 1
    Save 1.03 €
    2% Off
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Remel PathoDx and Extraction Reagent 2 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description PathoDx Extraction Reagent 2, 7mL/Vial
CE Marker Yes
Format Vial
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.