Learn More
Thermo Scientific™ Remel™ PathoDx™ Extraction Reagent 3
Product Code. 12781770
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
Product Code. Quantity Unit Size
12781770 Each Each
1 options
Product Code. 12781770 Supplier Thermo Scientific™ Supplier No. R62060
  • ONLINE EXCLUSIVE PRICE
    41.26 €
    38.88 € / Pack of 1
    Save 2.38 €
    5% Off
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Remel PathoDx and Extraction Reagent 3 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description PathoDx Extraction Reagent 3
CE Marker Yes
Quantity Each