Learn More
Rephile™ RephiDuo A Pack T1
Product Code. 15429739 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15429739 Water Purification System Each
1 options
Product Code. 15429739
  • ONLINE EXCLUSIVE PRICE
    417.48 €
    337.27 € / Pack of 1
    Save 80.21 €
    19% Off
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This product is specially designed to remove trace level ionic and organic contaminates from EDI, RO or distilled pure water to generate ultrapure water with high resistivity.
Rephile filter pack. Comparable to QGARDT1X1.
Compatible with following Millipore™ systems:
  • Milli-Q Advantage A10™
  • Milli-Q Reference™
Only compatible with systems manufactured before 2013.
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit