Learn More
Rephile™ RephiDuo A Pack T2
Product Code. 15439739 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15439739 Water Purification System Each
1 options
Product Code. 15439739
  • ONLINE EXCLUSIVE PRICE
    392.00 €
    337.27 € / Pack of 1
    Save 54.73 €
    13% Off
 Restricted Item
Estimated Shipment: 25-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This cartridge is specially designed to further purify Deionized (DI) water to produce high resistivity ultrapure water.
Rephile filter pack. Comparable to QGARDT2X1.
Compatible with following Millipore™ systems:
  • Milli-Q Advantage™
  • Milli-Q Reference™
Only compatible with systems manufactured before 2013.
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit