Learn More
Rephile™ RephiDuo L Pack L1
Product Code. 15469729 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15469729 Water Purification System Each
1 options
Product Code. 15469729
  • ONLINE EXCLUSIVE PRICE
    414.29 €
    334.21 € / Pack of 1
    Save 80.08 €
    19% Off
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This product is for additional/upstream pretreatment (extra protection) and designed to remove most of the particles, organic compounds and chlorine from tap water to protect the TL1 Pretreatment Pack and the RO membranes.
RephiDuo L Pack L1 for large Elix™ systems. Comparable to PRPK0L0S1.
Compatible with following Millipore™ systems:
  • Elix Clinical 35/70™
  • Elix 20/35/70/100™
  • RiOs 30/50/100/150/200™
  • Elix Gulfstream Clinical™
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit