Learn More
Rephile™ RephiDuo L Pack TL
Product Code. 15479729 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15479729 Water Purification System Each
1 options
Product Code. 15479729
  • ONLINE EXCLUSIVE PRICE
    440.00 €
    378.47 € / Pack of 1
    Save 61.53 €
    13% Off
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This product is designed to remove ionic and organic contaminants down to trace levels from pure water.
RephiDuo L Pack TL for large Elix™ systems. Comparable to QGARDTL01.
Compatible with following Millipore™ systems:
  • Elix Clinical 35/70™
  • Elix 20/35/70/100™
  • RiOs 30/50/100/150/200™
  • Elix Gulfstream Clinical™
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit