Learn More
Rephile™ RephiDuo L Pack TL1
Product Code. 15449729 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15449729 Water Purification System Each
1 options
Product Code. 15449729
  • ONLINE EXCLUSIVE PRICE
    369.00 €
    317.41 € / Pack of 1
    Save 51.59 €
    13% Off
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This product is for primary pretreatment (main RO protection) and designed to remove the most of the particles, organic compounds and chlorine from tap water to protect RO membranes.
Comparable to PR0GTL0S1.
Compatible with following Millipore™ systems:
  • Elix Clinical 35/70™
  • Elix 20/35/70/100™
  • RiOs 30/50/100/150/200™
  • Elix Gulfstream Clinical™
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit