Learn More
Rephile™ RephiDuo P Pack 1
Product Code. 15469719 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15469719 Water Purification System Each
1 options
Product Code. 15469719
  • ONLINE EXCLUSIVE PRICE
    428.13 €
    346.41 € / Pack of 1
    Save 81.72 €
    19% Off
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This product is designed to remove the most of the particles, organic compounds and chlorine from tap water to protect RO membranes.
RephiDuo P Pack 1 for Elix™ and Rios™ systems. Comparable to PR0G00001.
Compatible with following Millipore™ systems:
  • Direct-Q 3 /5™ (older version)
  • Elix (UV) 3/5/10™
  • RiOs 3™ (older version)
  • RiOs 5/8/16™
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit