Learn More
Wuhan Zhongze Trading Co Ltd™ RephiDuo P Pack S2
Product Code. 15419739 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Wuhan Zhongze Trading Co Ltd™ RephiDuo P S2
Product Code.
15419739 Wuhan Zhongze Trading Co Ltd™ RephiDuo P S2
1 options
Product Code. 15419739
  • ONLINE EXCLUSIVE PRICE
    383.40 €
    349.92 € / Pack of 1
    Save 33.48 €
    8% Off
Estimated Shipment: 15-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
RephiDuo P pack S2 for Elix and Rios systems.
RephiDuo P pack S2 for Elix and Rios systems. Comparable to MILPR0G0T0S2.
  • This product is designed to remove large particles and organic compounds, soften water, and remove chlorine from tap water to protect the RO membranes.
Specifications
Type Water Purification System