Learn More
Rephile™ RephiDuo U Pack 2
Product Code. 15429729 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15429729 Water Purification System Each
1 options
Product Code. 15429729
  • ONLINE EXCLUSIVE PRICE
    292.00 €
    250.93 € / Pack of 1
    Save 41.07 €
    14% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
This product is designed to remove ionic and organic contaminates down to trace levels from DI pure water.
RephiDuo U Pack 2 for MilliQ Century™ systems. Comparable to QGARD00D2.
Compatible with following Millipore™ systems:
  • Milli-Q Academic™
  • Milli-Q Biocel™
  • Milli-Q Element™
  • Milli-Q Gradient™
  • Milli-Q Synthesis™
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit