missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Replacement Battery

Product Code. 12479780 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
eLINE Electronic Pipettes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12479780 eLINE Electronic Pipettes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12479780 Supplier Sartorius Supplier No. 731009
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Replacement Battery for use with eLINE™ electronic pipets.

  • For use with: eLINE™ Electronic Pipettes
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Green
Disposable Reusable
For Use With Pipettes
Material Polypropylene
Product Line eLINE
Sterility Non-sterile
For Use With (Equipment) eLINE Electronic Pipettes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.