Learn More
Klever Replacement Blade
Product Code. 17768977
Change view
Click to view available options
Size:
Small
Unit Size:
Pack of 50
Product Code. Size Unit Size
17768977 Small Pack of 50
1 options
Product Code. 17768977 Supplier Klever Supplier No. HS8820SS
  • ONLINE EXCLUSIVE PRICE
    349.32 €
    300.86 € / Pack of 50
    Save 48.46 €
    13% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Klever replacement blade is a high-quality stainless steel blade, suitable for the safety knife KS-101SS-W.
  • High-quality stainless steel blade, suitable for the safety knife KS-101SS-W (100 pcs. in carton).
  • Can be used on both sides, therefore particularly economical.
Specifications
Blade Material Stainless Steel
Size Small
For Use With (Equipment) KS-101 SS Safety Cutters
Disposable Single-use
Sterility Non-sterile
Certifications/Compliance NSF
Quantity 10/Pk.