missing translation for 'onlineSavingsMsg'
Learn More

3M™ Replacement Clear Acetate Goggle Lens

Product Code. 15779807 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
3M™ Safety Goggles 2890A / 2890SAS
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15779807 3M™ Safety Goggles 2890A / 2890SAS Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15779807 Supplier 3M™ Supplier No. 7000061925
  • ONLINE EXCLUSIVE PRICE
    279.00 €
    190.35 € / Pack of 20
    Save 88.65 €
    31% Off
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M™ Replacement Clear Acetate Goggle Lens for use with 3M™ Safety Goggles 2890A / 2890SAS.

  • Replacement acetate lens for 3M™ Safety Goggles 2890A / 2890SAS
  • Increased durability with an extremely scratch resistant coating
TRUSTED_SUSTAINABILITY

Specifications

Lens Tint Clear
Product Type Replacement Lens
For Use With (Equipment) 3M™ Safety Goggles 2890A / 2890SAS
Lens Material Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.