Learn More
Klever Replacement Head (Wider Cut Head)
Product Code. 17789007
Change view
Click to view available options
Quantity:
12/Pk.
Unit Size:
Pack of 240
Product Code. Quantity Unit Size
17789007 12/Pk. Pack of 240
1 options
Product Code. 17789007 Supplier Klever Supplier No. KCJXH30
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Replacement Head (wider cut head) for use with Klever X-Change Safety Cutters.
  • '30' Blade head with wide mouth, therefore suitable for thicker materials.
  • For safety knives Klever XChange and Klever XChange Plus.
  • Especially safe thanks to the concealed blade.
Specifications
Product Type Replacement Head
For Use With (Equipment) Safety Knives Klever XChange and Klever XChange Plus
Disposable Single-use
Sterility Non-sterile
Quantity 12/Pk.