missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Replacement Items for Freezer/Mill™ Grinding Vial Sets

Product Code. 12578686 Shop All SPEX Certiprep Products
Change view
Click to view available options
Material:
Polycarbonate
Stainless Steel (440 C)
For Use With (Equipment):
6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set
6881 Vial Set
Spex SamplePrep Freezer/Mill Cryogenic Grinder
Unit Size:
Each
Pack of 2
Pack of 20
Pack of 4
Product Type:
Center Cylinder
End Plug
Replacement Impactor
Product Code. Material For Use With (Equipment) unitSize Product Type
12578686 Polycarbonate Spex SamplePrep Freezer/Mill Cryogenic Grinder Pack of 4 -
12508696 - - Each -
12558696 Polycarbonate 6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set Pack of 4 Center Cylinder
12578696 Polycarbonate Spex SamplePrep Freezer/Mill Cryogenic Grinder Pack of 20 Center Cylinder
12568696 Polycarbonate 6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set Pack of 20 Center Cylinder
15340482 Polycarbonate 6881 Vial Set Pack of 20 Center Cylinder
12588686 Stainless Steel (440 C) Spex SamplePrep Freezer/Mill Cryogenic Grinder Pack of 2 End Plug
12598686 Stainless Steel (440 C) Spex SamplePrep Freezer/Mill Cryogenic Grinder Each Replacement Impactor
8 options
This item is not returnable. View return policy

Product Code. 12578686

Brand: Cole-Parmer™ 6751C4

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Keep spare items to ensure there is no down time.

  • Suitable for use with the Cole-Parmer™ Freezer/Mill™ grinding vial sets
  • Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinding vial sets with a wide range of grinding ball options suitable for your application.

Specifications

For Use With (Equipment) Spex SamplePrep Freezer/Mill Cryogenic Grinder
Material Polycarbonate
Model Cole-Parmer Freezer/Mill
Packaging Quantity 4 Pk.
Sample Volume (English) 0.01 to 0.13 oz.
Sample Volume (Metric) 0.5 to 4 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.