Learn More
Thermo Scientific™ Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
Shop All Thermo Scientific Products
Click to view available options
Type:
ClickSeal Biocontainment Lid
O-ring
For Use With (Equipment):
MicroClick 24 x 2 Rotor
MicroClick 24 x 2 Rotor Lid
MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors
Unit Size:
Each
Description
A range of options for secure closure on microcentrifuge rotors to help contain samples.
- O-rings for inner and outer seals
- ClickSeal lids for optimal biocontainment
- Available for a variety of sizes and models
Specifications
Specifications
| Type | ClickSeal Biocontainment Lid |
| For Use With (Equipment) | MicroClick 24 x 2 Rotor |