Learn More
Fisherbrand™ Replacement Parts for MultiSUB Maxi Horizontal Gel System
Shop All Fisherbrand ProductsBeschreibung
MultiSUB Horizontal Gel Electrophoresis Systems combine economical buffer and gel consumption without compromising resolution and separation speed.
The large format of the MultiSUB Maxi enables the rapid separation of up to 550 samples. The MS Maxi is versatile enough for routine preparatory and high-throughput techniques, including RFLP analysis, southern and northern blotting applications, and DNA library and PCR fragment analysis. It is primarily designed for separating high numbers of samples from PCR or cloning.
- Electrophoresis Tank (Including Electrodes): A raised platform holds the UV sample trays, with side wells to hold buffer solution; connects to an external power source
- Asymmetrical Safety Lid: Notes the correct polarity and orientation of electrodes, with guides that enable the electrode thumb locators to click easily into the correct position
- Tank and lid are designed for use with 20 x 10cm, 20 x 15cm, 20 x 20cm or 20 x 25cm gel trays
Spezifikation
Spezifikation
| Color | Blue |
| For Use With (Application) | Nucleic Acid Gel Electrophoresis |
| For Use With (Equipment) | MultiSUB Maxi Horizontal Gel Electrophoresis Unit |
| Type | Lid |
| Gel Dimensions | 20 x 10 cm, 20 x 15 cm, 20 x 20 cm or 20 x 25 cm |
| Material | Molded Plastic |
| No. of Wells | For up to 550 separations |
| Quantity | 1 Each |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.