Learn More
KD Scientific Replacement Parts for Stainless Steel Syringes
Product Code. 17153541
Change view
Click to view available options
Type:
1/16 in. Swagelok Fitting
O-ring
Tip Seal O-Ring
For Use With:
100 mL Syringe
20 mL Syringe
20 to 200 mL Syringe
20 to 200 mL Syringes
200 mL Syringe
50 mL Syringe
Material:
Perfluoroelastomer
Stainless Steel
Viton
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Product Code. Type For Use With Material unitSize
17153541 O-ring 20 mL Syringe Perfluoroelastomer Pack of 2
15184214 O-ring 50 mL Syringe Perfluoroelastomer Pack of 2
17163541 O-ring 100 mL Syringe Perfluoroelastomer Pack of 2
17827571 O-ring 200 mL Syringe Perfluoroelastomer Pack of 2
15815465 O-ring 20 mL Syringe Viton Pack of 10
16164327 O-ring 100 mL Syringe Viton Pack of 10
17837571 O-ring 200 mL Syringe Viton Pack of 10
17173541 Tip Seal O-Ring 20 to 200 mL Syringe Perfluoroelastomer Each
16174327 Tip Seal O-Ring 20 to 200 mL Syringe Viton Pack of 20
15194224 1/16 in. Swagelok Fitting 20 to 200 mL Syringes Stainless Steel Each
10 options
Product Code. 17153541
Brand: KD Scientific 780834
Only null left
Add to basket
Add to basket
A well looked after stainless steel syringe can last a life time. KD Scientific provides replacement parts like O-rings, tip seals and various fittings to keep your syringes performing at the highest level.
  • Replacement seals
  • Perfluoroelastomer O-Rings
  • Perfluoroelastomer tip seal O-Rings
  • Viton O-Rings
  • Viton tip seal O-Rings
  • Swagelock™ fittings
  • Luer Lock Fittings
Specifications
For Use With 20 mL Syringe
IP Rating None
Material Perfluoroelastomer
Type O-ring