Learn More
Fisherbrand™ Replacement Probes for Traceable™ Conductivity and TDS Meters
Product Code. 11754216 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Model:
For 09-326-2 Meter
Unit Size:
Each
Product Code. Model Unit Size
11754216 For 09-326-2 Meter Each
1 options
Product Code. 11754216
  • ONLINE EXCLUSIVE PRICE
    301.00 €
    204.53 € / Pack of 1
    Save 96.47 €
    32% Off
View all promotions
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Traceable Conductivity and TDS Meters.
For use with Traceable Conductivity and TDS Meters.
Specifications
Connector Types BNC
For Use With (Equipment) Traceable Conductivity/TDS meter 09-326-2
Type Conductivity Probe
Material Glass
Measurement Ranges 0.05 to 200,000 μomhos
Model For 09-326-2 Meter