Learn More
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.
Brand: Lovibond™ 462910
Code : 04
Additional Details : Weight : 0.04100kg
Description
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.
- Highest quality production standards
- Consistent and reliable reagents are provided in liquid and tablet formats
- Maximum accuracy
Specifications
Hydrazine Test Powder | |
Water analysis | |
30 g |
Powder | |
Colorimeter and spectrophotometer | |
Comparator 2000+ replacement reagent |