missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.
52.90 € - 72.90 €
Specifications
For Use With (Application) | Water analysis |
---|---|
For Use With (Equipment) | Colorimeter and spectrophotometer |
Product Code | Brand | Format | Quantity | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Quantity | Type | Price | Quantity & Availability | |||
15331835
|
Lovibond™
515380BT |
Tablet | 100/Pk. | Iron HR Tablet |
56.80 €
Pack of 100 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
10747693
|
Lovibond™
513130BT |
Tablet | 100/Pk. | Silica No. 1 Tablet |
52.90 €
0.50kg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13111711
|
Lovibond™
462910 |
Powder | 30 g | Comparator 2000+ replacement reagent |
72.90 €
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||
15321715
|
Lovibond™
462910 |
Powder | 30 g | Comparator 2000+ replacement reagent |
72.90 €
30g |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
15311705
|
Lovibond™
537300 |
Powder | 100/Pk. | Persulfate Reagent Powder for CR |
65.40 €
Pack of 100 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
Description
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.
- Highest quality production standards
- Consistent and reliable reagents are provided in liquid and tablet formats
- Maximum accuracy
Specifications
Water analysis | |
Colorimeter and spectrophotometer |