Learn More
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.
Brand: Lovibond™ 537300
Code : NEW
Additional Details : CAS Number : 7727-21-1 Weight : 0.04500kg
Description
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.
- Highest quality production standards
- Consistent and reliable reagents are provided in liquid and tablet formats
- Maximum accuracy
Specifications
Powder | |
Colorimeter and spectrophotometer | |
Persulfate Reagent Powder for CR |
Water analysis | |
100/Pk. |
Safety and Handling
- persulphate reagent for CR
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H318-Causes serious eye damage.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : sodium hydrogensulphate, potassium persulphate