Learn More
Brady™ Repositionable Multipurpose Vinyl Cloth Labels
Easily place, remove and reposition labels on any clean, dry surface with the B-498 CleanLift™ series vinyl cloth label material. Print almost anywhere with a high-quality Brady™ handheld label printer.
Brand:  Brady™ 170789
Additional Details : Weight : 0.15900kg
 View more versions of this product
Product Code. 17900740
  • 53.85 € / Pack of 120
Estimated Shipment: 21-06-2024
to see stock.
Description
Description
  • Ideal for marking wires, cables, terminal blocks, wall plates
  • Constructed of tough coated vinyl cloth for resistance to extreme temperatures
  • Made with a strong rubber-based adhesive that can be cleanly removed and securely replaced on any clean, dry surface
Specifications
Specifications
UL Recognized to UL969, RoHS Compliant to 2005/618/EC MCV
Black on White
Die-cut
Labels
Barcode Labeling, Circuit Board Labeling, Component and Equipment Labeling, Laboratory Labeling, Wire and Cable Labeling
120 Labels/Cartridge
0.0073 in.
1.5 in.
2.54 cm
Repositionable Rubber
Semi-gloss
Vinyl Cloth
Non-sterile
BMP41, BMP51, M511
-40°F to +176°F
1.5 x 1 in.
1 in.