Learn More
Abcam Resazurin Assay Kit - Cell Cytotoxicity (Fluorometric)
Product Code. 32116076 Shop All Abcam Products
Change view
Click to view available options
Quantity:
5000 Tests
Unit Size:
5000 tests
Product Code. Quantity Unit Size
32116076 5000 Tests 5000 tests
1 options
Product Code. 32116076 Supplier Abcam Supplier No. ab1121195000Tests
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure 3H-Phenoxazin-3-one, 7-hydroxy-, 10-oxide, sodium salt in 1h in Cell Samples with Resazurin Assay Kit AB112119. Cited in 3 publications. Use with Microplate reader.
Measure 3H-Phenoxazin-3-one, 7-hydroxy-, 10-oxide, sodium salt in 1h in Cell Samples with Resazurin Assay Kit AB112119. Cited in 3 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 5000 Tests