missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Resazurin (Sodium Salt)

Product Code. 17110362
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17110362 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17110362 Supplier STEMCELL Technologies Supplier No. 75005
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cell proliferation and viability dye

  • Resazurin (7-hydroxy-10-oxidophenoxazin-10-ium-3-one, sodium) is a blue fluorogenic dye used as a redox indicator in cell viability and proliferation assays for bacteria, yeast or mammalian cells
  • The blue form of the dye is irreversibly reduced by enzymes in viable cells to generate a highly red-fluorescent product, resorufin, which exhibits an emission maximum at ∼595 nm and can be detected by flow cytometry, fluorescence microscopy, and high-throughput screening methods
  • Resazurin is minimally toxic to living cells, making it suitable for use in long-term cell culture
  • The dye has also been used to assay L-glutamate and to measure the metabolic activity of mitochondria

Specifications

CAS 62758-13-8
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.