missing translation for 'onlineSavingsMsg'
Learn More

Rescinnamine, TRC

Product Code. 30413642
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
Unit Size:
2.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30413642 5 mg 5mg
30410169 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30413642 Supplier Toronto Research Chemicals Supplier No. TRCR1447205MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24815-24-5
Molecular Formula C35 H42 N2 O9
Molecular Weight (g/mol) 634.72
Synonym Yohimban-16-carboxylic acid, 11,17-dimethoxy-18-[[(2E)-1-oxo-3-(3,4,5-trimethoxyphenyl)-2-propen-1-yl]oxy]-, methyl ester, (3β,16β,17α,18β,20α)-, 3β,20α-Yohimban-16β-carboxylic acid, 18β-hydroxy-11,17α-dimethoxy-, methyl ester, 3,4,5-trimethoxycinnamate (ester) (8CI), Rescinnamine (6CI), Yohimban-16-carboxylic acid, 11,17-dimethoxy-18-[[(2E)-1-oxo-3-(3,4,5-trimethoxyphenyl)-2-propenyl]oxy]-, methyl ester, (3β,16β,17α,18β,20α)- (9CI), Benz[g]indolo[2,3-a]quinolizine, yohimban-16-carboxylic acid deriv., 3,4,5-Trimethoxycinnamoyl methyl reserpate, Anaprel, Apoterin, Apoterin S, Cartric, Cinnaloid, Methyl trimethoxycinnamoylreserpate, Moderil, NSC 15628, O-(3,4,5-Trimethoxy-trans-cinnamoyl) methyl reserpate, Rescinnamin, Rescisan, Resealoid, Reserpic acid methyl ester 3,4,5-trimethoxycinnamate, Reserpinine, Reserpinine (C35 alkaloid), Scinnamina
IUPAC Name methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-[(E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoyl]oxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
SMILES CO[C@H]1[C@@H](C[C@@H]2CN3CCc4c([nH]c5cc(OC)ccc45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)\C=C\c6cc(OC)c(OC)c(OC)c6

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C35H42N2O9/c1-40-21-8-9-22-23-11-12-37-18-20-15-29(46-30(38)10-7-19-13-27(41-2)33(43-4)28(14-19)42-3)34(44-5)31(35(39)45-6)24(20)17-26(37)32(23)36-25(22)16-21/h7-10,13-14,16,20,24,26,29,31,34,36H,11-12,15,17-18H2,1-6H3/b10-7+/t20-,24+,26-,29-,31+,34+/m1/s1
Formula Weight 634.289
Chemical Name or Material Rescinnamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.