missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ resDNASEQ™ dPCR HEK293 DNA Kit

Product Code. 30054521 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30054521 100 reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30054521 Supplier Applied Biosystems™ Supplier No. A59365

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Applied Biosystems resDNASEQ dPCR HEK293 DNA Kit is a digital PCR (dPCR)-based solution to quantitate residual DNA from HEK293 expression systems, which are commonly used for production of gene therapies, cell-based vaccines, and similar therapeutics.

The Applied Biosystems resDNASEQ dPCR HEK293 DNA Kit is a digital PCR (dPCR)-based solution to quantitate residual DNA from HEK293 expression systems, which are commonly used for production of gene therapies, cell-based vaccines, and similar therapeutics. The resDNASEQ assay offers a fast and dependable method for accurately quantifying residual HEK293 DNA. This assay offers exceptional sensitivity and specificity, helping ensure that the quantitation data obtained from a wide range of sample types, including in-process samples with varying sample matrices and final product formulations, is reliable and consistent, instilling confidence in the results.

Features of the resDNASEQ dPCR HEK293 DNA Kit include:

  • Accurate, absolute quantitation of residual HEK293 DNA
  • Easy-to-use, integrated sample-to-results system features TaqMan dPCR master mix and TaqMan primer/probe set
  • Highly sensitive quantitation delivers results in about three hours
  • Optional manual or automated sample preparation, optimized for quantitative recovery of samples from complex matrices

Digital PCR workflow

The resDNASEQ dPCR HEK293 DNA Kit is part of an integrated dPCR workflow for impurity testing during biopharmaceutical manufacturing. Optional use of the Pharma KingFisher Apex 96 Deep-Well Magnetic Particle Processor with the PrepSEQ Nucleic Acid Sample Preparation Kit helps ensure high recovery of residual DNA with less labor and error than manual processing. The Pharma KingFisher Apex instrument can process up to 24 samples in triplicate, compared to three samples in triplicate using a manual method.

To help ensure performance that meets or exceeds regulatory compliance, the resDNASEQ kit has been internally validated on the QuantStudio Absolute Q Digital PCR System. Data analysis is streamlined using the QuantStudio Absolute Q dPCR Software, which includes accurate quantitation and security, audit, and e-signature capabilities to help enable 21 CFR Part 11 compliance.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Absolute Q Digital PCR Master Mix (5X), 0.36 mL, store at -15°C to -25°C before first use and 2°C to 8°C after first use, protect from light
• dPCR HEK293 DNA Assay Mix, 90 μL, store at -15°C to -25°C, protect from light
• Negative Control (water), 1.0 mL, store at -15°C to -25°C before first use and 2°C to 8°C after first use
Detection Method Digital PCR
Form Frozen
PCR Method Digital PCR
Label or Dye FAM + VIC
Product Line resDNASEQ
Quantity 100 reactions
Shipping Condition Dry Ice
Species HEK293
For Use With (Application) DNA Quantitation, Digital PCR (dPCR)

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.