missing translation for 'onlineSavingsMsg'
Learn More

Reserpine, 99%, Thermo Scientific Chemicals

Product Code. 11362186
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362186 5 g 5g
11352186 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362186 Supplier Thermo Scientific Alfa Aesar Supplier No. L03506.06
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50-55-5 | Molecular Formula: C33H40N2O9 | Molecular Weight: 608.688 g/mol

Inhibits vesicular uptake of catecholamines and serotoninReserpine is an antipsychotic and antihypertensive drug used to treat the hypertension, strokes, heart attacks and kidney problems. It is used to treat the symptoms of dyskinesia in patients who are affected by huntington’s disease. It is widely used as a reference material in the mass spectroscopy technique and also used as a sedative for horses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Incompatible with reducing agents and oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-55-5
Molecular Formula C33H40N2O9
Molecular Weight (g/mol) 608.688
MDL Number MFCD00005091
InChI Key QEVHRUUCFGRFIF-MDEJGZGSSA-N
Synonym reserpine, serpalan, serpasil, serpivite, apoplon, deserpine, elserpine, hiserpia, hypersil, raunervil
PubChem CID 5770
ChEBI CHEBI:28487
IUPAC Name methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-(3,4,5-trimethoxybenzoyl)oxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
SMILES COC1C(CC2CN3CCC4=C(C3CC2C1C(=O)OC)NC5=C4C=CC(=C5)OC)OC(=O)C6=CC(=C(C(=C6)OC)OC)OC

Specifications

Melting Point ∼265°C
Quantity 5 g
Beilstein 102014
Merck Index 14,8145
Solubility Information Soluble in water.
Formula Weight 608.69
Percent Purity 99%
Chemical Name or Material Reserpine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.