missing translation for 'onlineSavingsMsg'
Learn More

Reserpine, 99%, Thermo Scientific Chemicals

Product Code. p-7058274
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11352186

Brand: Thermo Scientific Alfa Aesar L03506.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits vesicular uptake of catecholamines and serotoninReserpine is an antipsychotic and antihypertensive drug used to treat the hypertension, strokes, heart attacks and kidney problems. It is used to treat the symptoms of dyskinesia in patients who are affected by huntington’s disease. It is widely used as a reference material in the mass spectroscopy technique and also used as a sedative for horses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Incompatible with reducing agents and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-55-5
Molecular Formula C33H40N2O9
Molecular Weight (g/mol) 608.688
MDL Number MFCD00005091
InChI Key QEVHRUUCFGRFIF-MDEJGZGSSA-N
Synonym reserpine, serpalan, serpasil, serpivite, apoplon, deserpine, elserpine, hiserpia, hypersil, raunervil
PubChem CID 5770
ChEBI CHEBI:28487
IUPAC Name methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-(3,4,5-trimethoxybenzoyl)oxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
SMILES COC1C(CC2CN3CCC4=C(C3CC2C1C(=O)OC)NC5=C4C=CC(=C5)OC)OC(=O)C6=CC(=C(C(=C6)OC)OC)OC

Specifications

Melting Point ∼265°C
Beilstein 102014
Merck Index 14,8145
Solubility Information Soluble in water.
Formula Weight 608.69
Percent Purity 99%
Quantity 1 g
Chemical Name or Material Reserpine
compliance-icons compliance-icons
Product Identifier
  • Reserpine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1A
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Reserpine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.