missing translation for 'onlineSavingsMsg'
Learn More

Reserpine, MedChemExpress

Product Code. 16499086
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499086 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499086 Supplier Medchem Express Supplier No. HYN0480100mg
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Reserpine is an inhibitor of the vesicular monoamine transporter 2 (VMAT2).

Reserpine is an inhibitor of the vesicular monoamine transporter 2 (VMAT2).

Specifications

Chemical Name or Material Reserpine
CAS 50-55-5
Color White
Purity Grade Notes Research
Molecular Formula C33H40N2O9
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (41.07 mM; Need ultrasonic)
SMILES O=C([C@H]([C@@H](OC)[C@H](OC(C1=CC(OC)=C(OC)C(OC)=C1)=O)C[C@]2([H])CN3CC4)[C@@]2([H])C[C@]3([H])C5=C4C(C=CC(OC)=C6)=C6N5)OC
Molecular Weight (g/mol) 608.68
Formula Weight 608.68
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.