missing translation for 'onlineSavingsMsg'
Learn More

Reserpine, TRC

Product Code. 30425039
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425039 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425039 Supplier Toronto Research Chemicals Supplier No. TRCR1446001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Reserpine
CAS 50-55-5
Molecular Formula C33 H40 N2 O9
Quantity 1 g
Synonym (3β,16β,17β,18β,20α)-11,17-Dimethoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]-yohimban-16-carboxylic acid methyl ester, 18β-Hydroxy-11,17β-dimethoxy-3β,20α-yohimban-16β-carboxylic acid methyl ester 3,4,5-trimethoxybenzoate (ester), 18β-Hydroxy-11,17β-dimethoxy-3β,20α-yohimban-16β-carboxylic acid methyl ester 3,4,5-trimethoxybenzoate, Benz[g]indolo[2,3-a]quinolizine','yohimban-16-carboxylic acid deriv., (-)-Reserpine, 11,17-Dimethoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]yohimban-16-carboxylic acid methyl ester, 3,4,5-Trimethoxybenzoyl methyl reserpate, Anquil, Apoplon, Banasil, Bioserpine, Carpacil, Crystoserpine, Deserpine, ENT 50146, Elserpine, Eserpine, Eskaserp, Hiposerpil, Kitine, Lemiserp, Loweserp, Mayserpine, Methyl (1S,2R,3R,4aS,13bR,14aS)-2,11-dimethoxy-3-[(3,4,5-trimethoxybenzoyl)oxy]-1,2,3,4,4a,5,7,8,13,13b,14,14a-dodecahydroindolo[2',3':3,4]pyrido[1,2-b]isoquinoline-1-carboxylate, Methyl reserpate 3,4,5-trimethoxybenzoate (ester), NSC 237659, NSC 59272, Quiescin, Rau-sed, Raucap, Raulen, Raunervil, Raupasil, Raurine, Rausedil, Rausedyl, Rausingle, Rauwasedin, Resercaps, Resercen, Reserlor, Reserpamed, Reserpex, Reserpic acid methyl ester 3,4,5-trimethoxybenzoate (ester), Reserpil, Reserpine, Reserpoid, Resine, Respital, Restran, Riserpa, Rivasin, Roxinoid, Sandril, Sedaraupin, Sedserp, Serfin, Serolfia, Serp-AFD, Serpaloid, Serpanray, Serpasil, Serpasil Serpasol, Serpasil premix, Serpasol, Serpate, Serpen, Serpentina, Serpicon, Serpiloid, Serpine, Serpine (pharmaceutical), Serpipur, Sertabs, Sertina, T-Serp, Temposerpine, Triserpin, Vio-Serpine
InChI Formula InChI=1S/C33H40N2O9/c1-38-19-7-8-20-21-9-10-35-16-18-13-27(44-32(36)17-11-25(39-2)30(41-4)26(12-17)40-3)31(42-5)28(33(37)43-6)22(18)15-24(35)29(21)34-23(20)14-19/h7-8,11-12,14,18,22,24,27-28,31,34H,9-10,13,15-16H2,1-6H3/t18-,22+,24-,27-,28+,31+/m1/s1
SMILES CO[C@H]1[C@@H](C[C@@H]2CN3CCc4c([nH]c5cc(OC)ccc45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)c6cc(OC)c(OC)c(OC)c6
IUPAC Name methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-(3,4,5-trimethoxybenzoyl)oxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
Molecular Weight (g/mol) 608.68
Formula Weight 608.2734
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.