missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Reservoir Bottles for Automatic Burette

Product Code. 10499261 Shop All Brand Products
Change view
Click to view available options
Color:
Amber
Clear
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10499261 Clear Each
10429641 Amber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10499261 Supplier BRAND™ Supplier No. 23310
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spare 2000mL bottles for automatic burette manufactured of boro.3.3 glass.

  • Manufactured of boro 3.3 glass with a PP screw-cap and PE-LD pouring ring
  • Maximum operating temperature is 80°C (cleaning temperatures should not exceed 60°C)
  • Requires use of bottletop adapter - 33mm/STJ 29/32
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Capacity (Metric) 2000 mL
For Use With (Equipment) Automatic Burette
Material Borosilicate Glass
Type Reservoir Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.