missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Residual Protein TrypLE™ ELISA Kit

Product Code. 30283273
Change view
Click to view available options
Quantity:
96 Tests kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30283273 96 Tests kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30283273 Supplier Gibco™ Supplier No. A6533101
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Residual Protein TrypLE ELISA Kit is a solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) for the quantitative detection of TrypLE enzyme.

The Residual Protein TrypLE ELISA Kit is a solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) for the quantitative detection of TrypLE enzyme. The kit is compatible with all forms of TrypLE enzyme, including TrypLE Select, TrypLE Express, and CTS TrypLE Select enzymes.

TrypLE enzyme is an animal-origin-free recombinant alternative to porcine trypsin. It is used for the dissociation of attachment-dependent cell lines. TrypLE enzyme dissociates cells cultured both in serum-free and serum-supplemented systems. Due to the unique biological sequence of TrypLE enzyme, traditional trypsin ELISA kits are ineffective at binding and detecting residual levels of TrypLE enzyme.

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.78 to 50 ng/mL
Assay Sensitivity 0.032 ng/mL
Content And Storage • Pre-coated 96-well plate
• Standard assay buffer concentrate
• HRP-conjugated detection antibody
• Wash buffer concentrate
• Chromogen stop solution
• Adhesive plate covers

Store at 2–8°C.

Conjugate Horseradish Peroxidase (HRP)
Immunoassay Kit Format Sandwich ELISA Kit
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 6.20%
Intraassay CV 7.20%
Label or Dye Horseradish Peroxidase (HRP)
Protein Aliases TrypLE, R-Protease
Quantity 96 Tests kit
Shipping Condition Wet Ice
Target TrypLE (R-Protease) concentration
Total Assay Time 1.5 hr.
Format Wash steps required
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • TRYPLE ELISA RESIDUAL KIT 1 EA
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 5-chloro-2-methylisothiazol-3(2H)-one, Phosphoric acid

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.