missing translation for 'onlineSavingsMsg'
Learn More

Resiniferatoxin, 99+%, Thermo Scientific™

Product Code. 15404839
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15404839 1 mg 1mg
15414839 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404839 Supplier Thermo Scientific Alfa Aesar Supplier No. J62458.M^
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Resiniferatoxin, a potent capsaicin-like stimulator of peripheral nociceptors in the neonatal rat tail in vitro. Resiniferatoxin is used as an anti-inflammatory, anti-viral and anti-psoriatic uses. Diterpene ester related to phorbol but apparently not tumorigenic activates protein kinase C

Solubility
Soluble in DMSO to 100mM and to 50mM in ethanol

Notes
Resiniferatoxin (RTX) is susceptible to slow degradation if exposed to air. Even at -20°C, Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep in a dry place. Keep away from strong oxidizing agents. RTX must be stored in an inert atmosphere for long-term stability.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 57444-62-9
Molecular Formula C37H40O9
Molecular Weight (g/mol) 628.72
MDL Number MFCD00135927,MFCD00135927
InChI Key OYWHJFQVSOUQSF-PKRQWDGKSA-N
Synonym resiniferatoxin
PubChem CID 57090162
IUPAC Name [(1R,2R,6R,10S,11R,15R,17R)-13-benzyl-6-hydroxy-4,17-dimethyl-5-oxo-15-(prop-1-en-2-yl)-12,14,18-trioxapentacyclo[11.4.1.0¹,¹⁰.0²,⁶.0¹¹,¹⁵]octadeca-3,8-dien-8-yl]methyl 2-(4-hydroxy-2-methoxyphenyl)acetate
SMILES COC1=CC(O)=CC=C1CC(=O)OCC1=C[C@H]2[C@H]3OC4(CC5=CC=CC=C5)O[C@]3(C[C@@H](C)[C@]2(O4)[C@@H]2C=C(C)C(=O)[C@@]2(O)C1)C(C)=C

Specifikationer

Color White
Quantity 1 mg
UN Number UN2811
Beilstein 5371150
Solubility Information Soluble in DMSO to 100mM and to 50mM in ethanol
Formula Weight 628.71
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material Resiniferatoxin
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.