missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Resiquimod

Product Code. 18055902
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18055902 50 mg 50mg
18005712 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18055902 Supplier R&D Systems™ Supplier No. 4536/50
  • ONLINE EXCLUSIVE PRICE
    540.20 €
    481.95 € / 50mg
    Save 58.25 €
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Toll-like receptor 7 (TLR7) agonist

Toll-like receptor 7 (TLR7) agonist. Induces upregulation of IL-6, IL-12, IFN-γ and iNOS expression in mouse bone marrow-derived macrophages (BMMs). Inhibits RANKL-induced osteoclast differentiation in mouse BMMs and human peripheral blood monocytes. Also promotes the differentiation of MDSCs into macrophages and dendritic cells. Proinflammatory and antiviral.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144875-48-9
Molecular Formula C17H22N4O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 314.38
Purity >98%
Molecular Weight (g/mol) 314.389
Chemical Name or Material Resiquimod
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.