missing translation for 'onlineSavingsMsg'
Learn More

Manutan Resist-A-Flame noticeboard, green, 1200mm x 2400mm

Product Code. 12838569
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12838569 Supplier Manutan Supplier No. 225M023NB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Open style noticeboard with the added safety of being manufactured with flame-retardant materials.
  • Green
  • Dimensions (H x W): 1200mm x 2400mm
  • Width (mm): 2400
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.