missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol-1,2,3-13C3, TRC

Product Code. 30420914
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420914 5 mg 5mg
30410043 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30420914 Supplier Toronto Research Chemicals Supplier No. TRCR1447025MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Resorcinol-1,2,3-13C3
CAS 1185048-67-2
Molecular Formula C313C3H6O2
Quantity 5 mg
Synonym 1,3-Benzenediol-13C3, 1,3-Dihydroxybenzene-13C3, 3-Hydroxyphenol-13C3, Fouramine RS-13C3, Fourrine 79-13C3, Rezorsine-13C3, Rodol RS-13C3, 1,3-Benzenediol-13C3, m-Phenylenediol-13C3
InChI Formula InChI=1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H/i4+1,5+1,6+1
SMILES O[13c]1[13cH][13c](O)ccc1
IUPAC Name (1,2,3-13C3)cyclohexa-1,3,5-triene-1,3-diol
Molecular Weight (g/mol) 113.09
Formula Weight 113.0468
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.