missing translation for 'onlineSavingsMsg'
Learn More

Cytiva RESOURCE™ Q IEX Columns

Product Code. 11564975 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
1 mL
6 mL
Unit Size:
1mL
6mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11564975 6 mL 6mL
10479944 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11564975 Supplier Cytiva Supplier No. 17117901
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Use for intermediate lab-scale purification and large-scale polishing of biomolecules Cytiva Life Sciences™ RESOURCE™ Q IEX Columns are strong anion exchanger columns.

  • Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics, sample loading of up to 25mg/mL and stability in organic solvents and pH extremes
  • Resource columns with 1mL bed volume have separation times of 3 minutes or less at 9.6mL/min. and approximately 20 min. using a peristaltic pump at 1mL/min.

TRUSTED_SUSTAINABILITY

Specifications

Description RESOURCE Q
Kit Contents Connectors for high- and low-pressure chromatography equipment and instructions
Column Material PEEK
Matrix Polystyrene/divinylbenzene
Flow Rate >60 mL/min.
For Use With (Application) RESOURCE Q columns are prepacked with SOURCE 15Q, a strong anion exchanger for high resolution polishing purification of proteins at high flow rates.
Ligand Type Quaternary ammonium
Content And Storage 4°C to 30°C in 20% Ethanol
Diameter (Metric) Inner Column 16 mm
Volume (Metric) 6 mL
Particle Size 15 μm
pH 2 to 12
Quantity 1 x 6 mL
Show More Show Less
compliance-icons
Product Identifier
  • RESOURCE Q, 6 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.