missing translation for 'onlineSavingsMsg'
Learn More

Cytiva RESOURCE™ S IEX Columns

Product Code. 10695565 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
1 mL
6 mL
Quantity:
1 x 1 mL
1 x 6 mL
Unit Size:
1mL
6mL
Product Code. Volume (Metric) Quantity unitSize
10695565 1 mL 1 x 1 mL 1mL
11574975 6 mL 1 x 6 mL 6mL
2 options
This item is not returnable. View return policy

Product Code. 10695565

Brand: Cytiva 17117801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

RESOURCE prepacked columns for fast separation. Resource S IEX Columns are prepacked with Source 15S, a strong cation exchanger for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.

  • Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics, sample loading of up to 25mg/mL and stability in organic solvents and pH extremes
  • Resource columns with 1mL bed volume have separation times of 3 minutes or less at 9.6mL/min. and approximately 20 min. using a peristaltic pump at 1mL/min.

TRUSTED_SUSTAINABILITY

Specifications

Description RESOURCE S
Kit Contents Connectors for high- and low-pressure chromatography equipment and instructions
Column Material PEEK
Matrix Polystyrene/divinylbenzene
Flow Rate >10 mL/min.
For Use With (Application) RESOURCE S columns are prepacked with SOURCE 15S, a strong cation exchanger for high-resolution polishing purification of proteins, at high flow rates.
Ligand Type Sulphonate group
Content And Storage 4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol
Diameter (Metric) Inner Column 6.4 mm
Volume (Metric) 1 mL
Particle Size 15 μm
pH 2 to 13
Quantity 1 x 1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.