missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Restore™ Fluorescent Western Blot Stripping Buffer

Product Code. 11835814 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 mL
100 mL
Unit Size:
100mL
20mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11835814 20 mL 20mL
11577900 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11835814 Supplier Thermo Scientific™ Supplier No. 62299

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Quickly and gently remove primary and near-infrared (IR) dye-labeled secondary antibodies from Western blots.

Thermo Scientific™ Restore Fluorescent Western Blot Stripping Buffer is a gentle and highly effective reagent for quickly removing primary and near-infrared (IR) dye-labeled secondary antibodies from Western blots.

Restore Fluorescent Western Blot Stripping Buffer enables the reuse of PVDF membranes, simplifying the Western blot optimization process and allowing the same blot to be reprobed with different primary antibodies to detect alternative targets. Restore Fluorescent Western Blot Stripping Buffer is for use with low-fluorescence PVDF membrane only (Part No. 22860).

Highlights:

  • Fast – strip blots in only 15 minutes at room temperature

  • Saves time – no need to run new gels and prepare a new blot

  • Conserve samples – reprobe the same PVDF membrane for multiple targets

  • Economical – less expensive than other commercially available stripping buffers

  • Efficient – effectively strips blots the first time

TRUSTED_SUSTAINABILITY

Specifications

Product Type Western Blot Stripping Buffer
Detection Method Fluorescence
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility PVDF
Product Line Restore
Quantity 20 mL
Content And Storage Upon receipt store at room temperature.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.