missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Resurge™ CD3 Supplement, without glucose and L-glutamine

Product Code. 16269911
Click to view available options
Quantity:
100 g
1 kg
5 kg
Unit Size:
100g
1kg
5kg
This item is not returnable. View return policy

Product Code. 16269911

Brand: Gibco™ 670020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A diverse set of fully chemically defined, animal origin–free and protein-free cell culture supplements, which help promote cell growth and boost protein production.

An animal origin-free (AOF) protein-free chemically defined supplement. It can be used in batch and fed batch mode in cell culture processes. All components are chemically defined with known molecular formulae and structures and meet our stringent Animal Origin-Free (AOF) specification. Formulated without glucose, L-glutamine, and sodium bicarbonate. Due to the balanced nutritional nature of this supplement, rich in amino acids, it has also been used in bacterial applications.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Cell Line CHO
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder
Product Type Media Supplement
Without Additives No Glutamine
Format Bottle(s)
Product Line Resurge
Quantity 5 kg
Source Animal origin-free
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.